Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253719.1 | NP_001240648.1 | 3 - 4 | 324 | 89 | |
NM_178673.4 | NP_848788.2 | 3 - 4 | 584 | 89 | ||
GenBank mRNA | AF374460.1 | - | 3 - 4 | 558 | 89 | |
AK033652.1 | - | 3 - 4 | 575 | 89 | ||
AK046070.1 | - | 3 - 4 | 325 | 89 | ||
AK081152.1 | - | 3 - 4 | 575 | 89 | ||
AK138388.1 | - | 3 - 4 | 876 | 89 | ||
AK140611.1 | - | 3 - 4 | 587 | 89 | ||
BC075640.1 | - | 3 - 4 | 547 | 30545529 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |