Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083901.1 | NP_001077370.1 | 8 - 9 | 1120 | 116 | |
NM_133680.3 | NP_598441.3 | 7 - 8 | 1217 | 116 | ||
GenBank mRNA | AK146596.1 | - | 8 - 9 | 1113 | 116 | |
BC033469.1 | - | 7 - 8 | 1179 | 4951631 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |