Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176968.4 | NP_795942.2 | 5 - 6 | 550 | 147 | |
GenBank mRNA | AK077876.1 | - | 5 - 6 | 551 | 147 | |
AK145666.1 | - | 5 - 6 | 551 | 147 | ||
AK161350.1 | - | 5 - 6 | 504 | 147 | ||
BC125276.1 | - | 5 - 6 | 549 | 40107511 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |