Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178035.4 | NP_828874.3 | 3 - 4 | 580 | 70 | |
GenBank mRNA | AB087503.1 | - | 3 - 4 | 548 | 70 | |
BC044804.1 | - | 3 - 4 | 438 | 70 | ||
BC132041.1 | - | 3 - 4 | 531 | 40130318 | 70 | |
BC132067.1 | - | 3 - 4 | 531 | 40130344 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |