Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178070.4 | NP_835171.2 | 9 - 10 | 1034 | 82 | |
GenBank mRNA | AK032076.1 | - | 9 - 10 | 1036 | 82 | |
AK134927.1 | - | 9 - 10 | 957 | 82 | ||
BC011059.1 | - | 9 - 10 | 1026 | 82 | ||
BC034170.1 | - | 9 - 10 | 746 | 4971695 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |