Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008312.4 | NP_032338.3 | 1 - 2 | 560 | 95 | |
GenBank mRNA | AK034512.1 | - | 1 - 2 | 562 | 95 | |
AK043320.1 | - | 1 - 2 | 561 | 95 | ||
AK079648.1 | - | 1 - 2 | 562 | 95 | ||
AK086348.1 | - | 1 - 2 | 559 | 95 | ||
AK139392.1 | - | 1 - 2 | 562 | 95 | ||
BC098327.1 | - | 1 - 2 | 541 | 95 | ||
BC141085.1 | - | 1 - 2 | 139 | 9055697 | 95 | |
M63685.1 | - | 1 - 2 | 423 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus 1700 Probe spans exons |