Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001288586.1 | NP_001275515.1 | 6 - 7 | 603 | 148 | |
NM_010786.4 | NP_034916.1 | 7 - 8 | 688 | 148 | ||
GenBank mRNA | AK004719.1 | - | 7 - 8 | 647 | 148 | |
AK088638.1 | - | 7 - 8 | 618 | 148 | ||
AK160069.1 | - | 6 - 7 | 479 | 148 | ||
BC050902.1 | - | 7 - 8 | 604 | 6415937 | 148 | |
BC092270.1 | - | 7 - 8 | 641 | 4018419 | 148 | |
U47934.1 | - | 5 - 6 | 394 | 148 | ||
X58876.1 | - | 7 - 8 | 595 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |