Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163552.1 | NP_001157024.1 | 7 - 8 | 1418 | 142 | |
NM_026193.2 | NP_080469.2 | 6 - 7 | 1631 | 142 | ||
GenBank mRNA | AK007733.1 | - | 6 - 7 | 1520 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |