Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnppd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026977.2 | NP_081253.1 | 7 - 8 | 904 | 65 | |
GenBank mRNA | AK029090.1 | - | 7 - 8 | 904 | 65 | |
AK080437.1 | - | 1 - 2 | 1122 | 65 | ||
AK145962.1 | - | 8 - 9 | 1151 | 65 | ||
AK159252.1 | - | 626 | 65 | |||
AK169779.1 | - | 7 - 8 | 1176 | 65 | ||
AK177413.1 | - | 3 - 4 | 226 | 65 | ||
BC013714.1 | - | 6 - 7 | 677 | 65 | ||
BC028815.1 | - | 7 - 8 | 889 | 5126467 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |