Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB030195.1 | - | 1 - 2 | 318 | 67 | |
AK014383.2 | - | 1 - 2 | 193 | 67 | ||
AK045045.1 | - | 1 - 2 | 327 | 67 | ||
BC103778.1 | - | 1 - 2 | 261 | 6493935 | 67 | |
BC116935.1 | - | 1 - 2 | 179 | 40126254 | 67 | |
BC116937.1 | - | 1 - 2 | 181 | 40126256 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |