Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025609.2 | NP_079885.2 | 7 - 8 | 776 | 93 | |
GenBank mRNA | AB088136.1 | - | 7 - 8 | 776 | 93 | |
BC027054.1 | - | 7 - 8 | 768 | 93 | ||
BC041110.1 | - | 7 - 8 | 771 | 93 | ||
BC054369.1 | - | 7 - 8 | 776 | 5356886 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |