Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009655.2 | NP_033785.1 | 9 - 10 | 1635 | 93 | |
GenBank mRNA | AB003026.1 | - | 9 - 10 | 1297 | 93 | |
AK030851.1 | - | 9 - 10 | 1636 | 93 | ||
AK031391.1 | - | 2711 | 93 | |||
BC027280.1 | - | 9 - 10 | 1292 | 3501215 | 93 | |
L25274.1 | - | 4 - 5 | 427 | 93 | ||
U95030.1 | - | 9 - 10 | 1103 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |