Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144953.1 | NP_001138425.1 | 9 - 10 | 1309 | 62 | |
NM_182636.4 | NP_872574.2 | 9 - 10 | 1309 | 62 | ||
GenBank mRNA | AK141799.1 | - | 9 - 10 | 1310 | 62 | |
BC052055.1 | - | 8 - 9 | 1255 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |