Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00009324.mm.1 |
NM_001102446.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009324.mm.1 |
NM_001102446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009324.mm.1 |
NM_009653.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009324.mm.1 |
NM_009653.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009324.mm.1 |
TC0X00001501.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00009323.mm.1 |
NM_009653.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009323.mm.1 |
NM_009653.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009323.mm.1 |
TC0X00001501.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00009319.mm.1 |
NM_001102446.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009319.mm.1 |
NM_001102446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009319.mm.1 |
NM_009653.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009319.mm.1 |
NM_009653.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009319.mm.1 |
TC0X00001501.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00009322.mm.1 |
NM_001102446.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009322.mm.1 |
NM_001102446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009322.mm.1 |
NM_009653.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009322.mm.1 |
NM_009653.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00009322.mm.1 |
TC0X00001501.mm.1 |
MTA TC |