Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080126.1 | NP_001073595.1 | 5 - 6 | 823 | 85 | |
NM_001277926.1 | NP_001264855.1 | 6 - 7 | 939 | 85 | ||
NM_009812.2 | NP_033942.1 | 5 - 6 | 996 | 85 | ||
GenBank mRNA | AF067834.1 | - | 5 - 6 | 876 | 85 | |
AJ000641.1 | - | 4 - 5 | 442 | 85 | ||
AK146758.1 | - | 5 - 6 | 996 | 85 | ||
AK153340.1 | - | 5 - 6 | 705 | 85 | ||
AK168812.1 | - | 5 - 6 | 996 | 85 | ||
BC006737.1 | - | 5 - 6 | 938 | 3964119 | 85 | |
BC049955.1 | - | 5 - 6 | 830 | 4208824 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |