Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010472.2 | NP_034602.1 | 4 - 5 | 774 | 61 | |
GenBank mRNA | AK029917.1 | - | 4 - 5 | 587 | 61 | |
AK048326.1 | - | 4 - 5 | 775 | 61 | ||
AK049400.1 | - | 4 - 5 | 761 | 61 | ||
BC031154.1 | - | 4 - 5 | 761 | 4976622 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |