Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026825.3 | NP_081101.3 | 26 - 27 | 2402 | 64 | |
GenBank mRNA | AK043591.1 | - | 11 - 12 | 982 | 64 | |
AK049059.1 | - | 8 - 9 | 778 | 64 | ||
AY437876.1 | - | 26 - 27 | 2407 | 64 | ||
BC012229.1 | - | 5 - 6 | 431 | 64 | ||
BC150819.1 | - | 25 - 26 | 2316 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |