Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026825.3 | NP_081101.3 | 20 - 21 | 1779 | 80 | |
GenBank mRNA | AK043591.1 | - | 5 - 6 | 359 | 80 | |
AK049059.1 | - | 2 - 3 | 155 | 80 | ||
AY437876.1 | - | 20 - 21 | 1784 | 80 | ||
BC150819.1 | - | 19 - 20 | 1693 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |