Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164743.1 | NP_001158215.1 | 6 - 7 | 972 | 93 | |
NM_134069.3 | NP_598830.2 | 7 - 8 | 1096 | 93 | ||
GenBank mRNA | AK128896.1 | - | 7 - 8 | 1080 | 93 | |
AK143896.1 | - | 7 - 8 | 1068 | 93 | ||
BC018252.1 | - | 7 - 8 | 1025 | 4240254 | 93 | |
BC024459.1 | - | 7 - 8 | 1052 | 4975341 | 93 | |
BC034179.1 | - | 7 - 8 | 916 | 4972500 | 93 | |
BC034183.1 | - | 7 - 8 | 901 | 4973620 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |