Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178640.2 | NP_848755.1 | 2 - 3 | 390 | 69 | |
GenBank mRNA | AB116655.1 | - | 2 - 3 | 259 | 69 | |
AK035259.1 | - | 2 - 3 | 391 | 69 | ||
AK041022.1 | - | 2 - 3 | 386 | 69 | ||
AK044785.1 | - | 2 - 3 | 444 | 69 | ||
AK084275.1 | - | 2 - 3 | 390 | 69 | ||
AK085691.1 | - | 2 - 3 | 391 | 69 | ||
AK134868.1 | - | 2 - 3 | 391 | 69 | ||
AK144880.1 | - | 2 - 3 | 221 | 69 | ||
AK151677.1 | - | 2 - 3 | 440 | 69 | ||
AK153362.1 | - | 2 - 3 | 440 | 69 | ||
AK167635.1 | - | 2 - 3 | 391 | 69 | ||
BC085110.1 | - | 2 - 3 | 399 | 30735904 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |