Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136065.2 | NP_001129537.1 | 5 - 6 | 1734 | 95 | |
NM_001294143.1 | NP_001281072.1 | 5 - 6 | 1810 | 95 | ||
NM_001294144.1 | NP_001281073.1 | 5 - 6 | 1810 | 95 | ||
NM_010433.2 | NP_034563.2 | 5 - 6 | 1810 | 95 | ||
GenBank mRNA | AF170301.1 | - | 5 - 6 | 1560 | 95 | |
AF170302.1 | - | 5 - 6 | 1560 | 95 | ||
AF208292.1 | - | 5 - 6 | 1647 | 95 | ||
AF273680.1 | - | 5 - 6 | 1531 | 95 | ||
AF333791.1 | - | 5 - 6 | 1437 | 95 | ||
AF333792.1 | - | 5 - 6 | 1437 | 95 | ||
BC031904.1 | - | 5 - 6 | 1543 | 5368577 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |