Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172772.2 | NP_766360.2 | 4 - 5 | 1178 | 97 | |
GenBank mRNA | AK035930.1 | - | 4 - 5 | 1179 | 97 | |
AK046403.1 | - | 4 - 5 | 1165 | 97 | ||
AK046623.1 | - | 4 - 5 | 1152 | 97 | ||
AK077470.1 | - | 1 - 2 | 110 | 97 | ||
AK220158.1 | - | 4 - 5 | 1207 | 97 | ||
BC049357.1 | - | 4 - 5 | 1155 | 97 | ||
BC055816.1 | - | 4 - 5 | 906 | 97 | ||
BC058683.1 | - | 4 - 5 | 1136 | 6834871 | 97 | |
BC069845.1 | - | 4 - 5 | 891 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |