Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027895.1 | NP_082171.1 | 10 - 11 | 1242 | 60 | |
GenBank mRNA | AK004783.1 | - | 8 - 9 | 1824 | 60 | |
AK032030.1 | - | 8 - 9 | 864 | 60 | ||
AK154642.1 | - | 10 - 11 | 1155 | 60 | ||
BC037093.1 | - | 10 - 11 | 1263 | 60 | ||
BC039958.1 | - | 6 - 7 | 604 | 60 | ||
BC151153.1 | - | 10 - 11 | 1168 | 9088047 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |