Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdxacb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198675.2 | NP_941077.2 | 1 - 2 | 219 | 70 | |
GenBank mRNA | AK085276.1 | - | 1 - 2 | 140 | 70 | |
AK135025.1 | - | 222 | 70 | |||
BY141558.1 | - | 1 - 2 | 221 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |