Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081037.2 | NP_001074506.1 | 4 - 5 | 611 | 80 | |
NM_001242411.1 | NP_001229340.1 | 4 - 5 | 611 | 80 | ||
GenBank mRNA | BC120892.1 | - | 4 - 5 | 604 | 40061454 | 80 |
BC120893.1 | - | 4 - 5 | 604 | 40061455 | 80 | |
CB521559.1 | - | 3 - 4 | 320 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |