Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164524.1 | NP_001157996.1 | 1 - 2 | 66 | 82 | |
GenBank mRNA | AK017139.1 | - | 1 - 2 | 55 | 82 | |
CN704519.1 | - | 1 - 2 | 66 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |