Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170966.1 | NP_001164437.1 | 4 - 5 | 595 | 135 | |
NM_001170967.1 | NP_001164438.1 | 4 - 5 | 595 | 135 | ||
NM_001170970.1 | NP_001164441.1 | 4 - 5 | 595 | 135 | ||
NM_028866.3 | NP_083142.2 | 4 - 5 | 595 | 135 | ||
GenBank mRNA | AB086191.1 | - | 3 - 4 | 374 | 135 | |
AK003256.1 | - | 4 - 5 | 563 | 135 | ||
AK009297.1 | - | 4 - 5 | 562 | 135 | ||
AK045125.1 | - | 4 - 5 | 555 | 135 | ||
AK045923.1 | - | 4 - 5 | 554 | 135 | ||
AK050653.1 | - | 4 - 5 | 555 | 135 | ||
AK161982.1 | - | 4 - 5 | 555 | 135 | ||
BC011156.1 | - | 4 - 5 | 507 | 135 | ||
BC019463.1 | - | 4 - 5 | 595 | 135 | ||
BC030076.1 | - | 4 - 5 | 527 | 135 | ||
BC064745.1 | - | 4 - 5 | 476 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |