Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000011807.mm.1 |
NM_001164237.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011807.mm.1 |
NM_001164237.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011807.mm.1 |
NM_026259.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011807.mm.1 |
NM_026259.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011807.mm.1 |
NR_104288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011807.mm.1 |
NR_104289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011807.mm.1 |
NR_104290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011807.mm.1 |
TC1000001605.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011809.mm.1 |
NM_001164237.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011809.mm.1 |
NM_001164237.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011809.mm.1 |
NR_104288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011809.mm.1 |
NR_104289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011809.mm.1 |
NR_104290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011809.mm.1 |
TC1000001605.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011808.mm.1 |
NM_001164237.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011808.mm.1 |
NM_001164237.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011808.mm.1 |
NR_104288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011808.mm.1 |
NR_104289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011808.mm.1 |
TC1000001605.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011806.mm.1 |
NM_001164237.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011806.mm.1 |
NM_001164237.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011806.mm.1 |
NM_026259.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011806.mm.1 |
NM_026259.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011806.mm.1 |
NR_104288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011806.mm.1 |
NR_104289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011806.mm.1 |
NR_104290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011806.mm.1 |
TC1000001605.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011811.mm.1 |
NM_001164237.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011811.mm.1 |
NM_001164237.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011811.mm.1 |
NR_104288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011811.mm.1 |
NR_104289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011811.mm.1 |
NR_104290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011811.mm.1 |
TC1000001605.mm.1 |
MTA TC |