Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177016.3 | NP_795990.2 | 3 - 4 | 400 | 104 | |
GenBank mRNA | AK033669.1 | - | 3 - 4 | 403 | 104 | |
AK162320.1 | - | 3 - 4 | 399 | 104 | ||
BC147415.1 | - | 3 - 4 | 361 | 9056491 | 104 | |
BC147416.1 | - | 3 - 4 | 361 | 9056498 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |