Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081219.2 | NP_001074688.1 | 9 - 10 | 893 | 68 | |
GenBank mRNA | AF009960.1 | - | 9 - 10 | 893 | 68 | |
BC147605.1 | - | 9 - 10 | 979 | 9056595 | 68 | |
BC147612.1 | - | 9 - 10 | 979 | 9056602 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |