Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252530.1 | NP_001239459.1 | 7 - 8 | 1290 | 92 | |
NM_001252531.1 | NP_001239460.1 | 7 - 8 | 1233 | 92 | ||
NM_175316.3 | NP_780525.1 | 7 - 8 | 1036 | 92 | ||
GenBank mRNA | AK041144.1 | - | 7 - 8 | 1232 | 92 | |
AK048120.1 | - | 7 - 8 | 1037 | 92 | ||
AK087682.1 | - | 7 - 8 | 1288 | 92 | ||
AK132394.1 | - | 7 - 8 | 1291 | 92 | ||
AK163542.1 | - | 7 - 8 | 1286 | 92 | ||
AK173049.1 | - | 5 - 6 | 822 | 92 | ||
BC019209.1 | - | 7 - 8 | 1019 | 92 | ||
BC037089.1 | - | 7 - 8 | 1008 | 92 | ||
BC049251.1 | - | 7 - 8 | 999 | 92 | ||
BC096485.1 | - | 7 - 8 | 1045 | 5101188 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |