Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033318.3 | NP_001028490.2 | 12 - 13 | 1768 | 81 | |
GenBank mRNA | AK030709.1 | - | 12 - 13 | 1735 | 81 | |
AK145163.1 | - | 12 - 13 | 1766 | 81 | ||
AK154693.1 | - | 12 - 13 | 1770 | 81 | ||
AK163143.1 | - | 1788 | 81 | |||
AK172200.1 | - | 12 - 13 | 1770 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |