Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177152.5 | NP_796126.4 | 10 - 11 | 1497 | 77 | |
GenBank mRNA | AK018522.1 | - | 4 - 5 | 310 | 77 | |
AK137007.1 | - | 10 - 11 | 1483 | 77 | ||
AY505341.1 | - | 10 - 11 | 1440 | 77 | ||
BC065142.1 | - | 10 - 11 | 1463 | 30610561 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |