Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010305.1 | NP_034435.1 | 5 - 6 | 838 | 96 | |
GenBank mRNA | BC032995.1 | - | 5 - 6 | 780 | 96 | |
BC138862.1 | - | 5 - 6 | 718 | 8861884 | 96 | |
BC138865.1 | - | 5 - 6 | 718 | 8861887 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |