Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016762.2 | NP_058042.2 | 7 - 8 | 1451 | 65 | |
GenBank mRNA | AK083151.1 | - | 7 - 8 | 1448 | 65 | |
AK138211.1 | - | 7 - 8 | 1448 | 65 | ||
AK163818.1 | - | 7 - 8 | 1410 | 65 | ||
BC005429.1 | - | 7 - 8 | 1321 | 3492881 | 65 | |
BC092298.1 | - | 7 - 8 | 1394 | 5063535 | 65 | |
U69262.1 | - | 7 - 8 | 1450 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |