Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177151.3 | NP_796125.2 | 46 - 47 | 8705 | 143 | |
GenBank mRNA | AK122302.1 | - | 7 - 8 | 1276 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |