Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172735.2 | NP_766323.1 | 5 - 6 | 628 | 81 | |
GenBank mRNA | AJ537495.1 | - | 5 - 6 | 620 | 81 | |
AK088527.1 | - | 5 - 6 | 629 | 81 | ||
AK146228.1 | - | 5 - 6 | 643 | 81 | ||
AK151694.1 | - | 5 - 6 | 624 | 81 | ||
AK167465.1 | - | 5 - 6 | 629 | 81 | ||
AK167992.1 | - | 5 - 6 | 643 | 81 | ||
AK168071.1 | - | 5 - 6 | 639 | 81 | ||
BC037445.1 | - | 5 - 6 | 582 | 81 | ||
BC050141.1 | - | 5 - 6 | 621 | 30011212 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |