Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081127.1 | NP_001074596.1 | 8 - 9 | 1509 | 65 | |
GenBank mRNA | AK145103.1 | - | 8 - 9 | 1511 | 65 | |
BC157953.1 | - | 8 - 9 | 1344 | 9007283 | 65 | |
BC158014.1 | - | 8 - 9 | 1342 | 9088151 | 65 | |
BC172106.1 | - | 8 - 9 | 1344 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |