Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsl1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025546.2 | NP_079822.1 | 3 - 4 | 417 | 130 | |
GenBank mRNA | AK012937.1 | - | 3 - 4 | 410 | 130 | |
AK075933.1 | - | 3 - 4 | 418 | 130 | ||
AK136025.1 | - | 3 - 4 | 384 | 130 | ||
AK145699.1 | - | 3 - 4 | 431 | 130 | ||
AK162241.1 | - | 3 - 4 | 391 | 130 | ||
AK165572.1 | - | 3 - 4 | 430 | 130 | ||
AK166579.1 | - | 3 - 4 | 413 | 130 | ||
AK168469.1 | - | 3 - 4 | 413 | 130 | ||
AK168717.1 | - | 3 - 4 | 406 | 130 | ||
AK171805.1 | - | 3 - 4 | 405 | 130 | ||
BC034355.1 | - | 3 - 4 | 383 | 2651674 | 130 | |
BC069934.1 | - | 3 - 4 | 391 | 130 | ||
DQ445383.1 | - | 3 - 4 | 425 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |