Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK090133.1 | - | 5 - 6 | 3766 | 66 | |
AK173328.1 | - | 2 - 3 | 294 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |