Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159394.1 | NP_001152866.1 | 4 - 5 | 649 | 56 | |
NM_001159395.1 | NP_001152867.1 | 5 - 6 | 531 | 56 | ||
NM_030612.3 | NP_085115.1 | 6 - 7 | 879 | 56 | ||
GenBank mRNA | AB020974.1 | - | 6 - 7 | 1091 | 56 | |
AB026551.2 | - | 4 - 5 | 649 | 56 | ||
AB047549.2 | - | 5 - 6 | 544 | 56 | ||
AK082908.1 | - | 4 - 5 | 648 | 56 | ||
AK136509.1 | - | 1 - 2 | 118 | 56 | ||
AK141318.1 | - | 4 - 5 | 648 | 56 | ||
AK149946.1 | - | 5 - 6 | 527 | 56 | ||
AK150519.1 | - | 4 - 5 | 648 | 56 | ||
AK152016.1 | - | 4 - 5 | 650 | 56 | ||
AK156826.1 | - | 4 - 5 | 650 | 56 | ||
AK159093.1 | - | 4 - 5 | 650 | 56 | ||
AK159171.1 | - | 4 - 5 | 650 | 56 | ||
AK159459.1 | - | 4 - 5 | 650 | 56 | ||
AK159645.1 | - | 4 - 5 | 650 | 56 | ||
AK159917.1 | - | 4 - 5 | 650 | 56 | ||
AK160065.1 | - | 4 - 5 | 648 | 56 | ||
BC058188.1 | - | 4 - 5 | 634 | 5341736 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |