Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037957.3 | NP_001033046.1 | 8 - 9 | 1345 | 157 | |
NM_001271370.1 | NP_001258299.1 | 8 - 9 | 1298 | 157 | ||
GenBank mRNA | AJ537610.1 | - | 8 - 9 | 1298 | 157 | |
BC019545.1 | - | 8 - 9 | 1345 | 4511845 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |