Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170907.1 | NP_001164378.1 | 1 - 2 | 240 | 122 | |
NM_019797.4 | NP_062771.2 | 4 - 5 | 1056 | 122 | ||
GenBank mRNA | AF197574.1 | - | 1 - 2 | 115 | 122 | |
AF539614.1 | - | 4 - 5 | 939 | 122 | ||
AK039024.1 | - | 1 - 2 | 125 | 122 | ||
AK078393.1 | - | 1 - 2 | 117 | 122 | ||
AK146298.1 | - | 1 - 2 | 123 | 122 | ||
AK150752.1 | - | 1 - 2 | 117 | 122 | ||
BC021316.1 | - | 1 - 2 | 128 | 5027899 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |