Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600003375.mm.1 |
NM_145933.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003375.mm.1 |
NM_145933.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003375.mm.1 |
TC1600000335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003379.mm.1 |
NM_001252505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003379.mm.1 |
NM_001252505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003379.mm.1 |
NM_145933.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003379.mm.1 |
NM_145933.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003379.mm.1 |
TC1600000335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003386.mm.1 |
NM_001252505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003386.mm.1 |
NM_001252505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003386.mm.1 |
NM_001252506.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003386.mm.1 |
NM_001252506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003386.mm.1 |
NM_145933.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003386.mm.1 |
NM_145933.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003386.mm.1 |
TC1600000335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003376.mm.1 |
NM_145933.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003376.mm.1 |
NM_145933.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003376.mm.1 |
TC1600000335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003378.mm.1 |
NM_001252505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003378.mm.1 |
NM_001252505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003378.mm.1 |
NM_145933.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003378.mm.1 |
NM_145933.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003378.mm.1 |
TC1600000335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003382.mm.1 |
NM_001252505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003382.mm.1 |
NM_001252505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003382.mm.1 |
NM_001252506.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003382.mm.1 |
NM_001252506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003382.mm.1 |
NM_145933.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003382.mm.1 |
NM_145933.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003382.mm.1 |
TC1600000335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003385.mm.1 |
NM_001252505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003385.mm.1 |
NM_001252505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003385.mm.1 |
NM_001252506.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003385.mm.1 |
NM_001252506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003385.mm.1 |
NM_145933.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003385.mm.1 |
NM_145933.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003385.mm.1 |
TC1600000335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003383.mm.1 |
NM_001252505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003383.mm.1 |
NM_001252505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003383.mm.1 |
NM_001252506.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003383.mm.1 |
NM_001252506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003383.mm.1 |
NM_145933.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003383.mm.1 |
NM_145933.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003383.mm.1 |
TC1600000335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003374.mm.1 |
NM_145933.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003374.mm.1 |
NM_145933.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003374.mm.1 |
TC1600000335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003377.mm.1 |
NM_001252505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003377.mm.1 |
NM_001252505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003377.mm.1 |
TC1600000335.mm.1 |
MTA TC |