Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081114.1 | NP_001074583.1 | 3 - 4 | 453 | 78 | |
GenBank mRNA | BC138413.1 | - | 2 - 3 | 372 | 8861433 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |