Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145152.4 | NP_660134.1 | 1 - 2 | 137 | 75 | |
GenBank mRNA | AK043535.1 | - | 1 - 2 | 112 | 75 | |
AY061858.1 | - | 1 - 2 | 110 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Probe spans exons |