Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177769.4 | NP_808437.2 | 3 - 4 | 524 | 83 | |
GenBank mRNA | AK029207.1 | - | 3 - 4 | 406 | 83 | |
AK132239.1 | - | 3 - 4 | 525 | 83 | ||
BC049160.1 | - | 3 - 4 | 387 | 83 | ||
BC120566.1 | - | 2 - 3 | 240 | 83 | ||
BC144961.1 | - | 2 - 3 | 240 | 83 | ||
BC144963.1 | - | 2 - 3 | 202 | 9053511 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |