Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019458.3 | NP_062331.2 | 4 - 5 | 504 | 77 | |
GenBank mRNA | AB041615.1 | - | 4 - 5 | 522 | 77 | |
AK017762.1 | - | 4 - 5 | 574 | 77 | ||
AK134857.1 | - | 4 - 5 | 517 | 77 | ||
AK148538.1 | - | 4 - 5 | 539 | 77 | ||
AK151665.1 | - | 4 - 5 | 550 | 77 | ||
AK152778.1 | - | 4 - 5 | 520 | 77 | ||
BC029843.1 | - | 4 - 5 | 504 | 2651081 | 77 | |
BC083337.1 | - | 4 - 5 | 485 | 5701517 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |