Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172787.2 | NP_766375.1 | 11 - 12 | 1188 | 79 | |
GenBank mRNA | AK045667.1 | - | 11 - 12 | 1189 | 79 | |
AK049597.1 | - | 11 - 12 | 1223 | 79 | ||
AK129443.1 | - | 7 - 8 | 733 | 79 | ||
BC082309.1 | - | 10 - 11 | 1058 | 30356672 | 79 | |
BC085192.1 | - | 10 - 11 | 1293 | 30694487 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |