Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005867.1 | NP_001005867.1 | 1 - 2 | 163 | 66 | |
NM_028601.3 | NP_082877.2 | 1 - 2 | 163 | 66 | ||
GenBank mRNA | BB645605.1 | - | 2 - 3 | 313 | 66 | |
BC060652.1 | - | 1 - 2 | 101 | 30354393 | 66 | |
CK617573.1 | - | 2 - 3 | 262 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |